Order Entry
Norway
ContactUsLinkComponent
Propargyl alcohol for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Health
Environment
Propargyl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
Corrosive
Health
Environment
Propargyl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1000 g/l (20 °C)
  • Melting Point:
    -53 °C
  • Molar Mass:
    56.06 g/mol
  • Boiling Point:
    114 - 115 °C
  • Vapor Pressure:
    10 hPa (20 °C)
  • Flash Point:
    33.5 °C
  • Refractive Index:
    1.4318 (20 °C, 589 nm)
  • Explosion Limit:
    1.9 - 86.2 %(V)
  • Density:
    0.95 g/cm3 (20 °C)
  • pH:
    3 (500 g/l, H2O, 20 °C)
  • Ignition Point:
    365 °C

Specification Test Results

Related Information