




Propargyl alcohol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-19-7
:
Formula:C₃H₄O
MDL Number:MFCD00002912
:56.06 g/mol
Merck Index:12,07993
Boiling Pt:114…115 °C (760 mmHg)
Melting Pt:–53 °C
Density:0.949 g/cm³
Flash Pt:34 °C
EINECS:203-471-2
UN:2927
ADR:6.1,II
:GHS06
:GHS05
:GHS09
:GHS02
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4300 to 1.4330 (20°C, 589 nm)
- GC:≥98.5 %