4-Chlorocinnamic acid
Glass bottle for solids, 25 g
Catalog # MOLEM29907936
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3-(4-Chlorophenyl)acrylic acid
- CAS Number:1615-02-7
- Formula:C₉H₇ClO₂
- MDL Number:MFCD00004396
- Molecular Weight:182.61 g/mol
- Boiling Pt:311 °C (1013 hPa)
- Melting Pt:252…254 °C
- EINECS:216-564-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



