Order Entry
Norway
ContactUsLinkComponent
Bisacrylamide ≥99%, white powder, Ultrapure
Irritant
Bisacrylamide ≥99%, white powder, Ultrapure
Katalog # ICNA04800172
Supplier:  MP Biomedicals
CAS Number:  
undefined
Irritant
Bisacrylamide ≥99%, white powder, Ultrapure
Katalog # ICNA04800172
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Synonyms:
    bis-Acrylamide; Methylenebisacrylamide; N,N'-Methylenediacrylamide; BIS
  • Form:
    White Powder
  • Storage:
    Store at room temperature (15 to 30 °C), protect from light, store under nitrogen
  • Conductivity:
    ≤10 µmhos
  • pH:
    ≥5 (2,5% aq soln)
  • Acrylic acid content:
    ≤0,02%

Specification Test Results

About this item

 :
 :154.17 g/mol
Boiling Pt:125 °C (3,33 hPa)
Melting Pt:185 °C
Density:1.235 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Ambient
EINECS:203-750-9
 :GHS07
CAS Number:110-26-9
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625