Benzilic acid
Catalog # MOLEM81489067
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:228.25 g/mol
Merck Index:13,01080
Boiling Pt:409 °C (1013 hPa)
Melting Pt:150…152 °C
Storage Temperature:Ambient
EINECS:200-993-2
CAS Number:76-93-7
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00004447