
(2,3-Dichlorophenyl)acetic acid 97%
ACRO365550250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:130 to 135 °C
- HPLC:≥96.0 %
:
:205.04 g/mol
:130…135 °C
:GHS07
:Ambient
:10236-60-9
:(C₆H₃Cl₂)CH₂COOH
:MFCD01861393