AMP (2-Amino-2-methylpropanol)
Catalog # MOLE41511752-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:13,00448
Boiling Pt:165 °C (1013 hPa)
Melting Pt:18…26 °C
Density:0.667 g/cm³ (20 °C)
Flash Pt:67 °C
:89.14 g/mol
Storage Temperature:Ambient
EINECS:204-709-8
:GHS07
CAS Number:124-68-5
Formula:(CH₃)₂C(NH₂)CH₂OH
MDL Number:MFCD00008051