Order Entry
Norway
ContactUsLinkComponent
Glycerine ≥99.0% (by GC), Reagent Grade
Glycerine ≥99.0% (by GC), Reagent Grade
Certificates
undefined
Glycerine ≥99.0% (by GC), Reagent Grade
Certificates

Specification Test Results

  • Vapor Density:
    3.1 (vs air)
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Water (Karl Fischer):
    Max. 1.0 %
  • Sulfated ash:
    Max. 0.1 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Vapor Pressure:
    <1 mmHg ( 20 °C)

Specification Test Results

Related Information