Order Entry
Norway
ContactUsLinkComponent
Ethanol for histology, J.T.Baker®
Ethanol for histology, J.T.Baker®
Katalog # 3405.9010
Supplier:  Avantor
CAS Number:  
Ethanol for histology, J.T.Baker®
Katalog # 3405.9010
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Assay (C2H5OH):
    min. 96 % (v/v)
  • Calcium (Ca):
    max. 5 ppm
  • Iron (Fe):
    max. 1 ppm
  • Residue after Evaporation:
    max. 0.0015 %
  • Substances Darkened by H2SO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0.001

Specification Test Results

About this item

 :
 :46.07 g/mol
Melting Pt:–114 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568