Order Entry
Norway
Orders LinkContactUsLinkComponent
2-Propanol

2-Propanol

208 L
Catalog # SWEDIP359902
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   SWED HANDLING
2-Propanol
Catalog # SWEDIP359902
Supplier:  SWED HANDLING
   Safer Human and Environmental Health
CAS Number:  67-63-0
2-Propanol
2-Propanol

208 L

CAS Number:  67-63-0
Price
kr 23 125,00
Log in for availability
1 * 208 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specifications

  • Pk:
    208 L
  • Cat. No.:
    SWEDIP359902
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.