

2-Propanol, EMPROVE® EXPERT Ph. Eur., BP, USP for preparative chromatography, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Sertifikat
Om dette produktet
CAS-nummer:67-63-0
:
Formel:(CH₃)₂CHOH
MDL-nummer:MFCD00011674
:60.1 g/mol
Merck-indeksen:13,05228
Kokende Pt:82 °C (1013 hPa)
Molten Pt:–89 °C
Tetthet:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Lagringstemperatur:Ambient
EINECS:200-661-7
U.N:1219
ADR:3,II
NÅ:01-2119457558-25
:GHS02
:GHS07
Spesifikasjoner
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion