1,2-Dimethoxyethane 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-71-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502
:90.12 g/mol
Merck Index:13,03251
Boiling Pt:84…86 °C (760 mmHg)
Melting Pt:–69 °C
Density:0.867 g/cm³
Flash Pt:–6 °C
Storage Temperature:Ambient
UN:2252
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:203-794-9
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3785 to 1.3815 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:40 to 100 ppm BHT
- Peroxide value:≤15 ppm