



Ethylenediamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-15-3
:
Formula:C₂H₈N₂
MDL Number:MFCD00008204
:60.1 g/mol
Merck Index:13,03829
Boiling Pt:117 °C (1013 hPa)
Melting Pt:8.5 °C
Density:0.898 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-468-6
UN:1604
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
Flash Pt:34 °C
:GHS02
:GHS06
:GHS05
:GHS08
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:11 °C
- Molar Mass:60.10 g/mol
- Boiling Point:116 °C (1013 hPa)
- Vapor Pressure:12 hPa (20 °C)
- Flash Point:38 °C
- Explosion Limit:2.5 - 16.3 %(V)
- Density:0.897 g/cm3 (20 °C)
- pH:12.2 (100 g/l, H2O, 20 °C)
- Ignition Point:405 °C
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.00947.0100_TDS_EN MERC8.00947.9051_TDS_EN ProductPage_800947
Product specification
msds_800947