Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)
Catalog # RESERES1161B-A103X
Supplier: RESEARCH ORGANICS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:209.24 g/mol
Melting Pt:102…103 °C
Storage Temperature:Ambient
EINECS:230-237-7
CAS Number:6976-37-0
Formula:C₈H₁₉NO₅
MDL Number:MFCD00002853