VWR®, Snood Caps
These disposable caps, with hairnet and peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Total hair covering, thanks to hairnet
- Available in white and blue
Weight: 10 g/m² (hairnet), 40 g/m² (top), 100 g/m² (peak)
: 100 pieces per bag, 10 bags per case.
- Catalog No:
- 113-8905
- 113-8904
- Garment application:
- Food
- Food