Hematin (from porcine blood) iron-containing porphyrin
Catalog # ABCAAB146273-1
Supplier: Abcam
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:633.49 g/mol
Storage Temperature:Refrigerator
CAS Number:15489-90-4
Formula:C₃₄H₃₃FeN₄O₅
MDL Number:MFCD00011615