Glysin USP, FCC, J.T.Baker®
: Avantor
:56-40-6
:
:NH₂CH₂COOH
:MFCD00008131
:75.07 g/mol
:13,04504
:181 °C (1013 hPa)
:83 °C
:1,64 g/cm³ (20 °C)
:Romtemperatur
:200-272-2
- Bulk Pharmaceutical Chemical:98.5 - 101.5 %
- USP - Assay (C₂H₅NO₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Loss on Drying:≤0.2 %
- USP - Residue on Ignition:≤0.1 %
- USP - Chloride (Cl):≤0.007 %
- USP - Sulfate (SO₄):≤0.0065 %
- USP - Hydrolyzable Substances:Passes Test
- USP - Related Compounds: Glycine anhydride:≤0.1 %
- USP - Related Compounds: Monochloroacetic acid (For Information Only):= 5 mg/kg
- USP - Related Compounds: Iminodiacetic acid:≤0.1 %
- USP - Related Compounds: Diglycine:≤0.1 %
- USP - Related Compounds: Triglycine:≤0.1 %
- USP - Related Compounds: Hexamethylenetetramine:≤0.1 %
- USP - Related Compounds: Total impurities:≤1.0 %
- FCC - Assay (C₂H₅NO₂) (dried basis):98.5 - 101.5 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤5 mg/kg
- FCC - Loss on Drying:≤0.2 %
- FCC - Residue on Ignition:≤0.1 %
- FCC - Insoluble Foreign Matter:Passes Test