Sodium acetate, anhydrous, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Merck Index:12,08711
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:365 g/l (20 °C)
- Melting Point:324 °C (decomposition)
- Molar Mass:82.04 g/mol
- Boiling Point:400 °C (decomposition)
- Vapor Pressure:(20 °C)
- Flash Point:250 °C
- Density:1.52 g/cm3 (20 °C)
- pH:7.5 - 9.0 (50 g/l, H2O, 20 °C)
- Ignition Point:607 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06268.0250_TDS_EN MERC1.06268.1000_TDS_EN ProductPage_106268