Sodium acetate, anhydrous ≥99.0%, crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Merck Index:12,08711
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
Specification Test Results
- Assay:≥99,0%
- pH (5% aq soln):7,0 - 9,2
- Insoluble Matter:≤0,01%
- Loss on Drying (120 °C):≤1,0%
- Chloride:≤0,002%
- Phosphate:≤0,001%
- Sulphate:≤0,003%
- Calcium:≤0,005%
- Magnesium:≤0,002%
- Heavy Metals (as Pb):≤0,001%
- Iron:≤0,001%