Butyl stearate
Catalog # EHERC16974300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Merck Index:13,01589
Boiling Pt:220 °C (1013 hPa)
Melting Pt:21 °C
Density:0.854 g/cm³ (20 °C)
Flash Pt:190 °C
:340.59 g/mol
Storage Temperature:Ambient
EINECS:204-666-5
:GHS07
CAS Number:123-95-5
Formula:C₂₂H₄₄O₂
MDL Number:MFCD00026669