Chemiluminescent acridinium esters
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and some peptides, and are frequently used as detection reagents in immunoassays.
- Species independent