Isopropyl formate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:625-55-8
:
Formula:C₄H₈O₂
MDL Number:MFCD00014127
:88.11 g/mol
Boiling Pt:68…71 °C
Melting Pt:–93 °C
Density:0.880
Flash Pt:–6 °C (21 °F)
Storage Temperature:Ambient
EINECS:210-901-2
UN:1281
ADR:3,II
:GHS02
:GHS07