kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Paracetamol
Paracetamol ≥98%
Paracetamol ≥98%
:
Thermo Fisher Scientific
Paracetamol ≥98%
:
Thermo Fisher Scientific
:
103-90-2
:
:
C₈H₉NO₂
:
MFCD00002328
:
151.16 g/mol
:
14,47
:
388 °C (1013 hPa)
:
168…172 °C
:
1.293
:
203-157-5
:
GHS07
:
Ambient
Appearance (Color):
White
Form:
crystalline powder
Assay (HPLC):
97.5%
msds_ALFAA11240
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years