Order Entry
Norway
Orders LinkContactUsLinkComponent
Formamide

Formamide

Glass bottle, 1 L
Catalog # MOLEM16396215
Health
Marketsource
Supplier:   Molekula
Formamide
Catalog # MOLEM16396215
Supplier:  Molekula
Marketsource
CAS Number:  75-12-7
Formamide
Formamide

Glass bottle, 1 L

CAS Number:  75-12-7
Price
kr 1 650,00
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:2 °C
  • Density:1.1334 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-842-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.