Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:78-93-3
:
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Merck Index:13,06097
Boiling Pt:79…81 °C
Melting Pt:–87 °C
Density:0.805 g/cm³ (20 °C)
Flash Pt:11.5 %(V)
Storage Temperature:Ambient
UN:1193
ADR:3,II
:72.11 g/mol
EINECS:201-159-0
REACH:01-2119457290-43
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Refractive Index:n20/D 1.379 (lit.)
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Non-volatile matter:Max. 0.0025 %
- Water (Karl Fischer):Max. 0.20 %
- Titrable Acid:Max. 0.0005 meq/g
- APHA:Max. 15
- Vapor Pressure:71 mmHg ( 20 °C)