


Dichloromethyl methyl ether 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4885-02-3
:
Formula:C₂H₄Cl₂O
MDL Number:MFCD00000828
:114.96 g/mol
Boiling Pt:85 °C (760 mmHg)
Density:1.27 g/cm³
Flash Pt:22 °C
EINECS:225-498-9
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4330 (20°C, 589 nm)
- GC:≥96.0 %