Order Entry
Norway
ContactUsLinkComponent
4-Bromo-1-butanol ≥80.0% (by NMR) (contains varying amounts of Tetrahydrofuran)
4-Bromo-1-butanol ≥80.0% (by NMR) (contains varying amounts of Tetrahydrofuran)
Supplier:  TCI
Certificates
undefined
4-Bromo-1-butanol ≥80.0% (by NMR) (contains varying amounts of Tetrahydrofuran)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (NMR):
    min. 80.0 w/w%

Specification Test Results