Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dibromobutyric acid
2,3-Dibromobutyric acid ≥97%
2,3-Dibromobutyric acid ≥97%
:
Thermo Fisher Scientific
2,3-Dibromobutyric acid ≥97%
:
Thermo Fisher Scientific
:
600-30-6
:
:
H₃C(BrCH)₂COOH
:
MFCD00014334
:
245.9 g/mol
:
100…110 °C (2 mmHg)
:
84…88 °C
:
Ambient
:
209-992-1
:
3261
:
8,III
:
GHS05
msds_ALFAL04841
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years