Acenaphthylene
Catalog # EHERC20510000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:152.2 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:91…92 °C
Density:0,8988 g/cm³ (20 °C)
EINECS:205-917-1
:GHS07
CAS Number:208-96-8
Formula:C₁₂H₈
MDL Number:MFCD00003806