L(-)-Tyrosine for synthesis, Sigma-Aldrich®
Catalog # 8.16021.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:0.38 g/l (20 °C)
- Melting Point:297 - 298 °C (decomposition)
- Molar Mass:181.19 g/mol
- Bulk Density:300 kg/m3
- Density:1.46 g/cm3 (20 °C)
- pH:6.5 (0.1 g/l, H2O)
Specification Test Results
About this item
:
:181.19 g/mol
Merck Index:13,09907
Boiling Pt:351 °C (1013 hPa)
Melting Pt:285 °C
Density:1.41 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-460-4
CAS Number:60-18-4
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606