Order Entry
Norway
Orders LinkContactUsLinkComponent
L(+)-Leucine USP
Glass bottle for solids, 250 g
  E811-250G
L(+)-Leucine USP
  E811-250G
 :  
 :  61-90-5
L(+)-Leucine USP
L(+)-Leucine USP
Glass bottle for solids, 250 g
 :  61-90-5
 :  (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
  •  :61-90-5
  •  :C₆H₁₃NO₂
  •  :MFCD00002617
  •  :131.17 g/mol
  •  :14,05451
  •  :300 °C
  •  :1.165 g/cm³ (20 °C)
  •  :Ambient
  •  :200-522-0
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g