Order Entry
Norway
Orders LinkContactUsLinkComponent
D-(-)-Mannitol, Millipore®

D-(-)-Mannitol, Millipore®

500 g
Catalog # 443907-500
D-(-)-Mannitol, Millipore®
Catalog # 443907-500
CAS Number:  69-65-8
D-(-)-Mannitol, Millipore®
D-(-)-Mannitol, Millipore®

500 g

CAS Number:  69-65-8
Price
kr 1 375,00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8

Documents

Specifications

Specifications

  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.