L(-)-Malic acid for racemat separation, for synthesis, Sigma-Aldrich®
: Merck
:97-67-6
:
:C₄H₆O₅
:MFCD00064213
:134.09 g/mol
:13,05730
:322 °C (1013 hPa)
:101…104 °C
:1.595 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:202-601-5
:GHS07
:Ambient
- Solubility:160 g/l (20 °C)
- Melting Point:101 - 104 °C
- Molar Mass:134.08 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:140 °C (decomposition)
- Density:1.60 g/cm3 (20 °C)
- pH:2.2 (10 g/l, H2O, 20 °C)