Organic reference standard, 2,3,3'-Trichlorobiphenyl (PCB No. 20) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-20
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:38444-84-7
- Description:2,3,3'-Trichlorobiphenyl (PCB No. 20) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,3,3'-Trichlorobiphenyl (PCB No. 20)