L(+)-Cysteine, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:52-90-4
:
Formula:HSCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064306
:121.16 g/mol
Merck Index:13,02810
Boiling Pt:254 °C (1013 hPa)
Melting Pt:115 °C
Density:1.523 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:200-158-2
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMERC1.02838.0025_TDS_EN ProductPage_102838
Product specification
msds_102838