Order Entry
Norway
ContactUsLinkComponent
L(+)-Cysteine, Millipore®
L(+)-Cysteine, Millipore®
Catalog # 243005-1
CAS Number:  
L(+)-Cysteine, Millipore®
Catalog # 243005-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :121.16 g/mol
Merck Index:13,02810
Boiling Pt:254 °C (1013 hPa)
Melting Pt:115 °C
Density:1.523 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-158-2
 :GHS07
CAS Number:52-90-4
Formula:HSCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064306