L(+)-Cysteine
Catalog # A3694.0100
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:121.16 g/mol
Merck Index:13,02810
Boiling Pt:254 °C (1013 hPa)
Melting Pt:115 °C
Density:1.523 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-158-2
:GHS07
CAS Number:52-90-4
Formula:HSCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064306