

Iron(III) nitrate nonahydrate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7782-61-8
:
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708
:404 g/mol
Merck Index:13,04057
Boiling Pt:125 °C (1013 hPa)
Melting Pt:47 °C
Density:1.68 g/cm³ (20 °C)
UN:1466
ADR:8,II
:M-Clarity™ quality level = MQ300
EINECS:233-899-5
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (complexometric):99.0 - 101.0 %
- Insoluble matter:≤ 0.005 %
- Free acid (as HNO₃):≤ 0.3 %
- Chloride (Cl):≤ 0.0005 %
- Phosphate (PO₄):≤ 0.005 %
- Sulfate (SO₄):≤ 0.005 %
- Ca (Calcium):≤ 0.005 %
- Cu (Copper):≤ 0.005 %
- K (Potassium):≤ 0.005 %
- Mg (Magnesium):≤ 0.001 %
- Mn (Manganese):≤ 0.02 %
- Na (Sodium):≤ 0.005 %
- Pb (Lead):≤ 0.001 %
- Zn (Zinc):≤ 0.001 %
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_103883
SDS
ProductPage_103883 ProductPage_103883