Iron(III) chloride hexahydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
:270.3 g/mol
Merck Index:13,04048
Boiling Pt:280…285 °C
Melting Pt:37 °C
Density:2.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
UN:3260
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay:Min. 99 %
- Free acid (as HCl):Max. 0.2 %
- Arsenic (As):Max. 0.0005 %
- Copper (Cu):Max. 0.002 %
- Iron (Fe 2⁺):Max. 0.005 %
- Lead (Pb):Max. 0.002 %
- Zinc (Zn):Max. 0.002 %
- Substances not precipitated by NH₄OH (SO₄):Max. 0.05 %
- Free chlorine (Cl):Max. 0.001 %
- Phosphate (PO₄):Max. 0.002 %
- Sulfate (SO₄):Max. 0.005 %
- Total N:Max. 0.001 %