Order Entry
Norway
ContactUsLinkComponent
Tin(II) chloride dihydrate
Tin(II) chloride dihydrate
Catalog # CAYM460107-1
Supplier:  Cayman Chemical
CAS Number:  
Tin(II) chloride dihydrate
Catalog # CAYM460107-1
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:10025-69-1
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863