Bole white (kaolin), Supelco®
Catalog # 1.04440.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
- Seq:4076
Specifications
About this item
:
:258.16 g/mol
Merck Index:13,05300
Melting Pt:1760 °C
Density:0.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:310-194-1
REACH:exempt
CAS Number:1332-58-7
Formula:Al₂SiO₇·2H₂O
MDL Number:MFCD00062311