Order Entry
Norway
ContactUsLinkComponent
Magnesium nitrate hexahydrate 99% ACS, Fluka™
Oxidizing
Magnesium nitrate hexahydrate 99% ACS, Fluka™
Certificates
Oxidizing
Magnesium nitrate hexahydrate 99% ACS, Fluka™

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • pH (5 %, 25 °C):
    5.0 - 8.2
  • Water insoluble matter:
    Max. 0.005 %
  • Barium (Ba):
    Max. 0.005 %
  • Calcium (Ca):
    Max. 0.01 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.005 %
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 0.005 %
  • Strontium (Sr):
    Max. 0.005 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.005 %
  • Ammonium (NH₄):
    Max. 0.003 %

Specification Test Results