Order Entry
Norway
ContactUsLinkComponent
Sodium tungstate dihydrate ≥99% ACS, Fluka™
Irriterende
Sodium tungstate dihydrate ≥99% ACS, Fluka™
Leverandør:  Honeywell Chemicals
Sertifikat
Irriterende
Sodium tungstate dihydrate ≥99% ACS, Fluka™
Leverandør:  Honeywell Chemicals
Sertifikat

Spesifikasjonstestresultater

  • Appearance:
    complying
  • Assay:
    99.0 - 101.0 %
  • Titrable Base:
    Max. 0.02 meq/g
  • Water insoluble matter:
    Max. 0.01 %
  • Molybdenum (Mo):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.005 %
  • Heavy metals (as Pb) and iron:
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.01 %

Spesifikasjonstestresultater

Relatert informasjon