
Potassium hexacyanoferrate(II) trihydrate 98.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14459-95-1
:
Formula:K₄Fe(CN)₆·3H₂O
MDL Number:MFCD00167023
:422.39 g/mol
Merck Index:12,07793
Melting Pt:70 °C
Density:1.85 g/cm³
EINECS:237-722-2
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Crystalline powder
- Titration with KMnO4:98.5 to 102.0 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):Passes test (limit about 0.01%)
- Insoluble matter:≤0.005 %