
2,4,6-Trihydroxybenzoic acid monohydrate ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71989-93-0
:
Formula:C₇H₆O₅·1H₂O
MDL Number:MFCD00149095
Melting Pt:∼ 210 °C (dec.)
:188.14 g/mol
EINECS:201-467-5
:GHS07
Storage Temperature:Ambient