Order Entry
Norway
ContactUsLinkComponent
 
 
 
EDTA disodium salt dihydrate 99-101% (by HPLC) Reag. Ph. Eur. for complexometry (NTA ≤0.1%), Fluka™
 
undefined
 
 
EDTA disodium salt dihydrate 99-101% (by HPLC) Reag. Ph. Eur. for complexometry (NTA ≤0.1%), Fluka™
 

 

  • Assay:
    99.0 - 101.0 %
  • Loss on drying (150°C, 3h):
    Max. 10 %
  • pH (5 %, 20°C):
    4.0 - 5
  • pH (5 %, 25 °C):
    4.0 - 6.0
  • Calcium (Ca):
    Max. 0.001 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Cobalt (Co):
    Max. 0.0005 %
  • Chromium (Cr):
    Max. 0.0005 %
  • Copper (Cu):
    Max. 0.0002 %
  • Iron (Fe):
    Max. 0.0005 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.0005 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Nitrilotriacetic acid (HPLC):
    Max. 0.1 %
  • Water insoluble matter:
    Max. 0.005 %
  • Appearance of the solution:
    complying
  • Identity (IR):
    complying