Zinc acetate dihydrate 98.0-101.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5970-45-6
:
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961
:219.51 g/mol
Merck Index:14,10128
Melting Pt:237 °C
Density:1.735
Storage Temperature:Ambient
EINECS:209-170-2
UN:3077
ADR:9,III
:GHS07
:GHS09