Guanidiniumtiocyanat
Catalog # MOLEM18456794
Supplier: Molekula
CAS Number:
Specifications
- Pakketype:Glass bottle for solids
- Fp:500 g
- Seq:3645
Specifications
About this item
:
:118.16 g/mol
Melting Pt:118 °C
Density:1,29 g/cm³ (20 °C)
EINECS:209-812-1
:GHS07
Storage Temperature:Romtemperatur
CAS Number:593-84-0
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027