Guanidinium thiocyanate for biotechnology
Supplier: Ambion
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
Melting Pt:118 °C
Density:1.29 g/cm³ (20 °C)
EINECS:209-812-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Abs.@280nm (70%, Water):< = 0.8
- Abs.@300nm (6M, Water):< 0.1
- Abs.@410nm (6M, Water):< 0.1
- Copper:< 0.0001 %
- DNase:none detected
- Solubility (70%, Water 50° C):pass
- Lead:< 0.0001 %
- Melting Point:118 °C - 121 °C
- Protease:none detected
- Purity:> 99 %
- RNase:none detected