Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Guanidiniumklorid
Guanidiniumklorid
Guanidiniumklorid
Guanidiniumklorid
RESERES5134G-A102X
:
RESEARCH ORGANICS
:
Guanidiniumklorid
RESERES5134G-A102X
:
RESEARCH ORGANICS
:
Pakketype:
Glass bottle for solids
Fp:
500 g
Seq:
3645
:
:
95.53 g/mol
:
13,04578
:
185 °C
:
1,354 g/cm³ (20 °C)
:
200-002-3
:
GHS07
:
Romtemperatur
:
50-01-1
:
NH₂C(=NH)NH₂·HCl
:
MFCD00013026